Anti-LAMP2b Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB18529-100EA 4327.94 DKK
ABCAAB18529-100
Anti-LAMP2b Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LAMP2b
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR