Anti-IL21 Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB18496-50EA 5430 DKK
ABCAAB18496-50
Anti-IL21 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: IL21 (interleukin 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR