Anti-FOXK1 Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB18196-100EA 3874.89 DKK
ABCAAB18196-100
Anti-FOXK1 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: FOXK1 (forkhead box K1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human