Anti-CXCL5 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB18134-50EA 4098.45 DKK
ABCAAB18134-50
Anti-CXCL5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CXCL5 (chemokine (C-X-C motif) ligand 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR