Anti-L Glutamate Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB18127-25EA 3953.69 DKK
ABCAAB18127-25
Anti-L Glutamate Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: L Glutamate
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human