Anti-Plasma Cell Mouse Monoclonal Antibody [clone: SPM310]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB18096-7EA 5630 DKK
ABCAAB18096-7
Anti-Plasma Cell Mouse Monoclonal Antibody [clone: SPM310]
Antistoffer
Type: Primary
Antigen: Plasma Cell
Clonality: monoclonal
Clone: SPM310
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR