Anti-FEN1 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17994-100EA 5800 DKK
ABCAAB17994-100
Anti-FEN1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: FEN1 (flap structure-specific endonuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR