Anti-Neurofibromin Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17963-50EA 5800 DKK
ABCAAB17963-50
Anti-Neurofibromin Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Neurofibromin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR