Anti-ERBB2 Mouse monoclonal antibody [clone: SPM172]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17897EA 4360 DKK
ABCAAB17897
Anti-ERBB2 Mouse monoclonal antibody [clone: SPM172]
Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: monoclonal
Clone: SPM172
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR