Anti-SARM1 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17812-50EA 3954.84 DKK
ABCAAB17812-50
Anti-SARM1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SARM1 (sterile alpha and TIR motif containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR