Anti-TYMP Mouse monoclonal antibody [clone: SPM322]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17806-7EA 4360 DKK
ABCAAB17806-7
Anti-TYMP Mouse monoclonal antibody [clone: SPM322]
Type: Primary
Antigen: TYMP (thymidine phosphorylase)
Clonality: monoclonal
Clone: SPM322
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR