Anti-ERCC4 Mouse monoclonal antibody [clone: SPM228]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17798-500EA 4610 DKK
ABCAAB17798-500
Anti-ERCC4 Mouse monoclonal antibody [clone: SPM228]
Type: Primary
Antigen: ERCC4 (excision repair cross-complementation group 4)
Clonality: monoclonal
Clone: SPM228
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR