Anti-Macrophage Inflammatory Protein 3 Goat polyclonal antibody Biotin

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17714-25EA 4140 DKK
ABCAAB17714-25
Anti-Macrophage Inflammatory Protein 3 Goat polyclonal antibody Biotin
Type: Primary
Antigen: Macrophage Inflammatory Protein 3
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR