Anti-SAA@ Mouse monoclonal antibody [clone: 3H73]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17466-50EA 4300 DKK
ABCAAB17466-50
Anti-SAA@ Mouse monoclonal antibody [clone: 3H73]
Type: Primary
Antigen: SAA@ (serum amyloid A1 cluster)
Clonality: monoclonal
Clone: 3H73
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR