Anti-C3 Mouse monoclonal antibody unconjugated [clone: 12E2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17456-100EA 5570 DKK
ABCAAB17456-100
Anti-C3 Mouse monoclonal antibody unconjugated [clone: 12E2]
Antistoffer
Type: Primary
Antigen: C3 (complement component 3)
Clonality: monoclonal
Clone: 1200
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR