Anti-SNCB Rabbit polyclonal antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB17394-500EA 4300 DKK
ABCAAB17394-500
Anti-SNCB Rabbit polyclonal antibody
Type: Primary
Antigen: SNCB (synuclein, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat