Anti-TGF beta Receptor II Sheep Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17282-100EA 5380 DKK
ABCAAB17282-100
Anti-TGF beta Receptor II Sheep Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TGF beta Receptor II
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Chicken
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR