Anti-F2 Mouse monoclonal antibody unconjugated [clone: 5G9]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17199-100EA 5880 DKK
ABCAAB17199-100
Anti-F2 Mouse monoclonal antibody unconjugated [clone: 5G9]
Antistoffer
Type: Primary
Antigen: F2 (coagulation factor II (thrombin))
Clonality: monoclonal
Clone: 5G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR