Anti-HPV16 E2 Mouse Monoclonal Antibody [clone: TVG 271]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17190-50EA 5630 DKK
ABCAAB17190-50
Anti-HPV16 E2 Mouse Monoclonal Antibody [clone: TVG 271]
Antistoffer
Type: Primary
Antigen: HPV16 E2
Clonality: monoclonal
Clone: TVG 271
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR