Anti-ARIH2 Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17014-100EA 5370 DKK
ABCAAB17014-100
Anti-ARIH2 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ARIH2 (ariadne RBR E3 ubiquitin protein ligase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR