Anti-BRF2 Goat polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB17011-100EA 5630 DKK
ABCAAB17011-100
Anti-BRF2 Goat polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: BRF2 (BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR