Anti-TLR2 Mouse monoclonal antibody Biotin [clone: mT2.7]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16908-25EA 4650 DKK
ABCAAB16908-25
Anti-TLR2 Mouse monoclonal antibody Biotin [clone: mT2.7]
Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: mT2.7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR