Anti-CASP9 Mouse monoclonal antibody [clone: 1-2]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16893-50EA 4060 DKK
ABCAAB16893-50
Anti-CASP9 Mouse monoclonal antibody [clone: 1-2]
Type: Primary
Antigen: CASP9 (caspase 9, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: 42006
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR