Anti-CSPS Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16870-100EA 5430 DKK
ABCAAB16870-100
Anti-CSPS Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: CSPS (catecholamine-sulfating phenol sulfotransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR