Anti-ESR2 Mouse monoclonal antibody [clone: 9.88]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16813-100EA 4250 DKK
ABCAAB16813-100
Anti-ESR2 Mouse monoclonal antibody [clone: 9.88]
Type: Primary
Antigen: ESR2 (estrogen receptor 2 (ER beta))
Clonality: monoclonal
Clone: 9.88
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR