Anti-DCC Mouse monoclonal antibody [clone: AF5]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16793-50EA 4350 DKK
ABCAAB16793-50
Anti-DCC Mouse monoclonal antibody [clone: AF5]
Type: Primary
Antigen: DCC (deleted in colorectal carcinoma)
Clonality: monoclonal
Clone: AF5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR