Anti-TP53 Mouse monoclonal antibody [clone: PAb1620]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16776-100EA 4360 DKK
ABCAAB16776-100
Anti-TP53 Mouse monoclonal antibody [clone: PAb1620]
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: monoclonal
Clone: PAb1620
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR