Anti-PRDX4 Mouse monoclonal antibody [clone: 1A1]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16754-100EA 4990 DKK
ABCAAB16754-100
Anti-PRDX4 Mouse monoclonal antibody [clone: 1A1]
Type: Primary
Antigen: PRDX4 (peroxiredoxin 4)
Clonality: monoclonal
Clone: 1A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR