Anti-PRDX3 Mouse monoclonal antibody [clone: 2B11]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16752-100EA 4690 DKK
ABCAAB16752-100
Anti-PRDX3 Mouse monoclonal antibody [clone: 2B11]
Type: Primary
Antigen: PRDX3 (peroxiredoxin 3)
Clonality: monoclonal
Clone: 2B11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR