Anti-CACNB3 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16717-100EA 5760 DKK
ABCAAB16717-100
Anti-CACNB3 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: CACNB3 (calcium channel, voltage-dependent, beta 3 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR