Anti-ESR1 Rabbit Monoclonal Antibody [clone: SP1]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16660-250EA 6280 DKK
ABCAAB16660-250
Anti-ESR1 Rabbit Monoclonal Antibody [clone: SP1]
Antistoffer
Type: Primary
Antigen: ESR1 (estrogen receptor 1)
Clonality: monoclonal
Clone: SP1
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR