Anti-AES Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16638-100EA 4360 DKK
ABCAAB16638-100
Anti-AES Rabbit polyclonal antibody
Type: Primary
Antigen: AES (amino-terminal enhancer of split)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR