Anti-Phosphotyrosine Mouse Monoclonal Antibody (Biotin) [clone: PY20]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16388-100EA 5630 DKK
ABCAAB16388-100
Anti-Phosphotyrosine Mouse Monoclonal Antibody (Biotin) [clone: PY20]
Antistoffer
Type: Primary
Antigen: Phosphotyrosine
Clonality: monoclonal
Clone: PY20
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR