Anti-IL13 Rat monoclonal antibody [clone: JES10-5A2]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16219-250EA 4460 DKK
ABCAAB16219-250
Anti-IL13 Rat monoclonal antibody [clone: JES10-5A2]
Type: Primary
Antigen: IL13 (interleukin 13)
Clonality: monoclonal
Clone: JES10-5A2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR