Anti-VIPR1 Mouse monoclonal antibody [clone: AS58]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16155-50EA 4360 DKK
ABCAAB16155-50
Anti-VIPR1 Mouse monoclonal antibody [clone: AS58]
Type: Primary
Antigen: VIPR1 (vasoactive intestinal peptide receptor 1)
Clonality: monoclonal
Clone: AS58
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR