Anti-Prostaglandin E Receptor EP2 Mouse Monoclonal Antibody [clone: 3E6]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16151-50EA 5670 DKK
ABCAAB16151-50
Anti-Prostaglandin E Receptor EP2 Mouse Monoclonal Antibody [clone: 3E6]
Antistoffer
Type: Primary
Antigen: Prostaglandin E Receptor EP2
Clonality: monoclonal
Clone: 3000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR