Anti-VSX2 Sheep Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16141-250EA 5910 DKK
ABCAAB16141-250
Anti-VSX2 Sheep Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: VSX2 (visual system homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR