Anti-CLU Mouse Monoclonal Antibody [clone: CLI-9]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB16077-50EA 5620 DKK
ABCAAB16077-50
Anti-CLU Mouse Monoclonal Antibody [clone: CLI-9]
Antistoffer
Type: Primary
Antigen: CLU (clusterin)
Clonality: monoclonal
Clone: CLI-9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR