Anti-GABARAPL2 Chicken polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15858-50EA 4360 DKK
ABCAAB15858-50
Anti-GABARAPL2 Chicken polyclonal antibody
Type: Primary
Antigen: GABARAPL2 (GABA(A) receptor-associated protein-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR