Anti-CPLX1 Chicken Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15855-50EA 5880 DKK
ABCAAB15855-50
Anti-CPLX1 Chicken Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CPLX1 (complexin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR