Anti-V5 Tag Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB15828-100EA 3651.34 DKK
ABCAAB15828-100
Anti-V5 Tag Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: V5 Tag (V5 tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu