Anti-FHL2 Goat Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB15715-100EA 5370 DKK
ABCAAB15715-100
Anti-FHL2 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: FHL2 (four and a half LIM domains 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human