Anti-CPT1B Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15703-100EA 5670 DKK
ABCAAB15703-100
Anti-CPT1B Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CPT1B (carnitine palmitoyltransferase 1B (muscle))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR