Anti-SFTPD Mouse Monoclonal Antibody (Biotin) [clone: VIF11]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15696-100EA 5450 DKK
ABCAAB15696-100
Anti-SFTPD Mouse Monoclonal Antibody (Biotin) [clone: VIF11]
Antistoffer
Type: Primary
Antigen: SFTPD (surfactant protein D)
Clonality: monoclonal
Clone: VIF11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR