Anti-GRM2 Mouse Monoclonal Antibody [clone: mG2Na-s]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15672-50EA 5600 DKK
ABCAAB15672-50
Anti-GRM2 Mouse Monoclonal Antibody [clone: mG2Na-s]
Antistoffer
Type: Primary
Antigen: GRM2 (glutamate receptor, metabotropic 2)
Clonality: monoclonal
Clone: mG2Na-s
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR