Anti-SNCB Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15533-7EA 4690 DKK
ABCAAB15533-7
Anti-SNCB Rabbit polyclonal antibody
Type: Primary
Antigen: SNCB (synuclein, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR