Anti-STAT5A Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15525-7EA 4690 DKK
ABCAAB15525-7
Anti-STAT5A Rabbit polyclonal antibody
Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR