Anti-NOS2 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15323-500EA 4104.08 DKK
ABCAAB15323-500
Anti-NOS2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NOS2 (nitric oxide synthase 2, inducible)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR