Anti-Ionotropic glutamate receptor 2 + 3 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15302-7EA 0 DKK
ABCAAB15302-7
Anti-Ionotropic glutamate receptor 2 + 3 Rabbit polyclonal antibody
Type: Primary
Antigen: Ionotropic glutamate receptor 2 + 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR