Anti-CLDN5 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15106-500EA 6010 DKK
ABCAAB15106-500
Anti-CLDN5 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: CLDN5 (claudin 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR